Learn More
Biomatik Corporation NCOA5 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08106
Encompass_Preferred
Ncoa5 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NCOA5. The Ncoa5 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nuclear Receptor Coactivator 5 Protein (112-283Aa). Applications: ELISA, WB, IF. Target Synonyms: NCOA5. Accession Number: Q9HCD5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7690