Learn More
Biomatik Corporation PEMT anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08108
Encompass_Preferred
Pemt Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PEMT. The Pemt Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phosphatidylethanolamine N-Methyltransferase Protein (2-49Aa). Applications: ELISA, WB, IF. Target Synonyms: PEMT. Accession Number: Q9UBM1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7692