Learn More
Biomatik Corporation ATXN2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08114
Encompass_Preferred
The ATXN2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Ataxin-2 protein (174-383AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:2000-1:5000, IF:1:50-1:200. Alternative Names: Ataxin-2, Spinocerebellar ataxia type 2 protein, Trinucleotide repeat-containing gene 13 protein, ATXN2, ATX2, SCA2, TNRC13. UniProt: Q99700. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7698