Learn More
Biomatik Corporation PARD6G anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08115
Encompass_Preferred
Pard6G Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PARD6G. The Pard6G Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Partitioning Defective 6 Homolog Gamma Protein (249-326Aa). Applications: ELISA, WB, IF. Target Synonyms: PARD6G. Accession Number: Q9BYG4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7699