Learn More
Biomatik Corporation IQCG anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08116
Encompass_Preferred
Iqcg Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IQCG. The Iqcg Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Iq Domain-Containing Protein G Protein (1-134Aa). Applications: ELISA, WB, IF. Target Synonyms: IQCG. Accession Number: Q9H095. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7700