Learn More
Biomatik Corporation ZC4H2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08117
Encompass_Preferred
Zc4H2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZC4H2. The Zc4H2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Zinc Finger C4H2 Domain-Containing Protein (1-224Aa). Applications: ELISA, WB, IF. Target Synonyms: ZC4H2. Accession Number: Q9NQZ6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7701