Learn More
Biomatik Corporation ALDH1A3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08120
Encompass_Preferred
The ALDH1A3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Aldehyde dehydrogenase family 1 member A3 protein (2-210AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:2000-1:10000, IF:1:50-1:200. Alternative Names: Aldehyde dehydrogenase family 1 member A3, Aldehyde dehydrogenase 6, Retinaldehyde dehydrogenase 3, RALDH-3, RalDH3, ALDH1A3, ALDH6. UniProt: P47895. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7704