Learn More
Biomatik Corporation FGFR1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08126
Encompass_Preferred
The FGFR1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Fibroblast growth factor receptor 1 protein (22-96AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:2000-1:10000, IF:1:50-1:200. Alternative Names: Fibroblast growth factor receptor 1, Basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, Fms-like tyrosine kinase 2, FLT-2, N-sam, Proto-oncogene c-Fg, FGFR1, BFGFR, CEK, FGFBR, FLG, FLT2, HBGFR. UniProt: P11362. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7710