Learn More
Biomatik Corporation CNN2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08136
Encompass_Preferred
The CNN2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Calponin-2 protein (2-309AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:2000-1:10000, IF:1:50-1:200. Alternative Names: Calponin-2, Calponin H2, smooth muscle, Neutral calponin, CNN2. UniProt: Q99439. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7720