Learn More
Biomatik Corporation SCIN anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507728
Encompass_Preferred
Scin Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SCIN. The Scin Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Adseverin Protein (556-703Aa). Applications: ELISA, WB, IF. Target Synonyms: SCIN. Accession Number: Q9Y6U3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7728 Supplier Biomatik Corporation Supplier No. CAC08144
Scin Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SCIN. The Scin Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Adseverin Protein (556-703Aa). Applications: ELISA, WB, IF. Target Synonyms: SCIN. Accession Number: Q9Y6U3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.