Learn More
Biomatik Corporation CDH5 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08147
Encompass_Preferred
The CDH5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Cadherin-5 protein (349-569AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Cadherin-5, 7B4 antigen, Vascular endothelial cadherin, VE-cadherin, CD144, CDH5. UniProt: P33151. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7731