Learn More
Biomatik Corporation LRP8 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08149
Encompass_Preferred
Lrp8 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LRP8. The Lrp8 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Low-Density Lipoprotein Receptor-Related Protein 8 Protein (131-332Aa). Applications: ELISA, WB, IF. Target Synonyms: LRP8. Accession Number: Q14114. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7733