Learn More
Biomatik Corporation FOXE1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08156
Encompass_Preferred
The FOXE1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Forkhead box protein E1 protein (184-365AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Forkhead box protein E1, Forkhead box protein E2, Forkhead-related protein FKHL15, HFKH4, HNF-3/fork head-like protein 5, HFKL5, Thyroid transcription factor 2, TTF-2, FOXE1, FKHL15, FOXE2, TITF2, TTF2. UniProt: O00358. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7740