Learn More
Biomatik Corporation IFI44L anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08158
Encompass_Preferred
Ifi44L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFI44L. The Ifi44L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Induced Protein 44-Like Protein (206-307Aa). Applications: ELISA, WB, IF. Target Synonyms: IFI44L. Accession Number: Q53G44. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7742