Learn More
Biomatik Corporation DST anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08167
Encompass_Preferred
The DST Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Dystonin protein (1-195AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Dystonin, 230 kDa bullous pemphigoid antigen, 230/240 kDa bullous pemphigoid antigen, Bullous pemphigoid antigen 1, BPA, Bullous pemphigoid antigen, Dystonia musculorum protein, Hemidesmosomal plaque protein, DST, BP230, BP240, BPAG1, DMH, DT, KIAA0728. UniProt: Q03001. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7751