Learn More
Biomatik Corporation FHL1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08171
Encompass_Preferred
The FHL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human Four and a half LIM domains protein 1 protein (13-240AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Four and a half LIM domains protein 1, FHL-1, Skeletal muscle LIM-protein 1, SLIM, SLIM-1, FHL1, SLIM1. UniProt: Q13642. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7755