Learn More
Biomatik Corporation FSIP2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08172
Encompass_Preferred
The FSIP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Fibrous sheath-interacting protein 2 protein (5530-5824AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Fibrous sheath-interacting protein 2, FSIP2. UniProt: Q5CZC0. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7756