Learn More
Biomatik Corporation DNAH12 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507758
Encompass_Preferred
The DNAH12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Dynein heavy chain 12, axonemal protein (2423-2632AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Dynein heavy chain 12, axonemal, Axonemal beta dynein heavy chain 12, Axonemal dynein heavy chain 12-like protein, Axonemal dynein heavy chain 7-like protein, Ciliary dynein heavy chain 12, Dynein heavy chain 7-like, axonemal, Dynein heavy chain domain-containing protein 2, DNAH12, DHC3, DLP12, DNAH12L, DNAH7L, DNAHC3, DNHD2, HDHC3, HL19. UniProt: Q6ZR08. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7758 Supplier Biomatik Corporation Supplier No. CAC08174
The DNAH12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Dynein heavy chain 12, axonemal protein (2423-2632AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Dynein heavy chain 12, axonemal, Axonemal beta dynein heavy chain 12, Axonemal dynein heavy chain 12-like protein, Axonemal dynein heavy chain 7-like protein, Ciliary dynein heavy chain 12, Dynein heavy chain 7-like, axonemal, Dynein heavy chain domain-containing protein 2, DNAH12, DHC3, DLP12, DNAH12L, DNAH7L, DNAHC3, DNHD2, HDHC3, HL19. UniProt: Q6ZR08. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.