Learn More
Biomatik Corporation CPEB2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08175
Encompass_Preferred
The CPEB2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Cytoplasmic polyadenylation element-binding protein 2 protein (167-308AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Cytoplasmic polyadenylation element-binding protein 2, CPE-BP2, CPE-binding protein 2, hCPEB-2, CPEB2. UniProt: Q7Z5Q1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7759