Learn More
Biomatik Corporation C16orf62 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507760
Encompass_Preferred
The C16orf62 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human UPF0505 protein C16orf62 protein (6-205AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: UPF0505 protein C16orf62, Esophageal cancer-associated protein, C16orf62, 101F10.2. UniProt: Q7Z3J2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7760 Supplier Biomatik Corporation Supplier No. CAC08176
The C16orf62 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human UPF0505 protein C16orf62 protein (6-205AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: UPF0505 protein C16orf62, Esophageal cancer-associated protein, C16orf62, 101F10.2. UniProt: Q7Z3J2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.