Learn More
Biomatik Corporation GULP1 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08182
Encompass_Preferred
The GULP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human PTB domain-containing engulfment adapter protein 1 protein (199-279AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:2000-1:10000, IF:1:50-1:500. Alternative Names: PTB domain-containing engulfment adapter protein 1, Cell death protein 6 homolog, PTB domain adapter protein CED-6, Protein GULP, GULP1, CED6, GULP. UniProt: Q9UBP9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7766