Learn More
Biomatik Corporation NPL anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507769
Encompass_Preferred
Npl Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NPL. The Npl Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human N-Acetylneuraminate Lyase Protein (17-77Aa). Applications: ELISA, WB, IF. Target Synonyms: NPL. Accession Number: Q9BXD5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7769 Supplier Biomatik Corporation Supplier No. CAC08185
Npl Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NPL. The Npl Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human N-Acetylneuraminate Lyase Protein (17-77Aa). Applications: ELISA, WB, IF. Target Synonyms: NPL. Accession Number: Q9BXD5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.