Learn More
Biomatik Corporation SOCS7 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08197
Encompass_Preferred
Socs7 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SOCS7. The Socs7 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Suppressor Of Cytokine Signaling 7 Protein (6-165Aa). Applications: ELISA, WB, IF. Target Synonyms: SOCS7. Accession Number: O14512. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7781