Learn More
Biomatik Corporation MAPRE2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08198
Encompass_Preferred
Mapre2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MAPRE2. The Mapre2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Microtubule-Associated Protein Rp; Eb Family Member 2 Protein (159-240Aa). Applications: ELISA, WB, IF. Target Synonyms: MAPRE2. Accession Number: Q15555. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7782