Learn More
Biomatik Corporation FOXI1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08199
Encompass_Preferred
The FOXI1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant Human Forkhead box protein I1 protein (183-343AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:500-1:5000, IF:1:50-1:200. Alternative Names: Forkhead box protein I1, Forkhead-related protein FKHL10, Forkhead-related transcription factor 6, FREAC-6, Hepatocyte nuclear factor 3 forkhead homolog 3, HFH-3, HNF-3/fork-head homolog 3, FOXI1, FKHL10, FREAC6. UniProt: Q12951. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7783