Learn More
Biomatik Corporation ARHGDIA anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08202
Encompass_Preferred
The ARHGDIA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Rho GDP-dissociation inhibitor 1 protein (2-204AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Rho GDI 1, Rho-GDI alpha, ARHGDIA, GDIA1. UniProt: P52565. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7786