Learn More
Biomatik Corporation AADACL2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08203
Encompass_Preferred
The AADACL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Arylacetamide deacetylase-like 2 protein (181-379AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Arylacetamide deacetylase-like 2, AADACL2. UniProt: Q6P093. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7787