Learn More
Biomatik Corporation ABHD2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507788
Encompass_Preferred
The ABHD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Abhydrolase domain-containing protein 2 protein (31-320AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Abhydrolase domain-containing protein 2 (EC:3.1.1.-), ABHD2, LABH2, Lung alpha/beta hydrolase 2, Protein PHPS1-2. UniProt: P08910. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7788 Supplier Biomatik Corporation Supplier No. CAC08204
The ABHD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Abhydrolase domain-containing protein 2 protein (31-320AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Abhydrolase domain-containing protein 2 (EC:3.1.1.-), ABHD2, LABH2, Lung alpha/beta hydrolase 2, Protein PHPS1-2. UniProt: P08910. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.