Learn More
Biomatik Corporation ACLY anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08207
Encompass_Preferred
The ACLY Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ATP-citrate synthase protein (4-265AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: ATP-citrate synthase, ATP-citrate (pro-S-)-lyase, ACL, Citrate cleavage enzyme, ACLY. UniProt: P53396. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7791