Learn More
Biomatik Corporation ACTRT2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08208
Encompass_Preferred
The ACTRT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Actin-related protein T2 protein (1-376AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Actin-related protein T2, ARP-T2, ACTRT2, ARPM2, Actin-related protein M2. UniProt: Q8TDY3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7792