Learn More
Biomatik Corporation ADA anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08209
Encompass_Preferred
The ADA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Adenosine deaminase protein (157-343AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Adenosine deaminase, Adenosine aminohydrolase, ADA, ADA1. UniProt: P00813. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7793