Learn More
Biomatik Corporation ATP6V1G1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507796
Encompass_Preferred
The ATP6V1G1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human V-type proton ATPase subunit G 1 protein (2-118AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: V-ATPase 13 kDa subunit 1; Vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16. UniProt: O75348. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7796 Supplier Biomatik Corporation Supplier No. CAC08212
The ATP6V1G1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human V-type proton ATPase subunit G 1 protein (2-118AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: V-ATPase 13 kDa subunit 1; Vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16. UniProt: O75348. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.