Learn More
Biomatik Corporation ADM anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08214
Encompass_Preferred
The ADM Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ADM protein (45-92AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: ProAM N-terminal 20 peptide, PAMP, ProAM-N20. UniProt: P35318. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7798