Learn More
Biomatik Corporation ADORA2A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08215
Encompass_Preferred
The ADORA2A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Adenosine receptor A2a protein (291-412AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Adenosine receptor A2a, ADORA2A, ADORA2. UniProt: P29274. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7799