Learn More
Biomatik Corporation AGA anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507800
Encompass_Preferred
The AGA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human N (4)- (beta-N-acetylglucosaminyl)-L-asparaginase protein (206-346AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Aspartylglucosaminidase, Glycosylasparaginase, N4- (N-acetyl-beta-glucosaminyl)-L-asparagine amidase, AGA. UniProt: P20933. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7800 Supplier Biomatik Corporation Supplier No. CAC08216
The AGA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human N (4)- (beta-N-acetylglucosaminyl)-L-asparaginase protein (206-346AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Aspartylglucosaminidase, Glycosylasparaginase, N4- (N-acetyl-beta-glucosaminyl)-L-asparagine amidase, AGA. UniProt: P20933. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.