Learn More
Biomatik Corporation Aif1 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08219
Encompass_Preferred
The Aif1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse Allograft inflammatory factor 1 protein (2-147AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: onized calcium-binding adapter molecule 1, Iba1, Aif1. UniProt: O70200. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7803