Learn More
Biomatik Corporation AKR1C2 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08221
Encompass_Preferred
The AKR1C2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Aldo-keto reductase family 1 member C2 protein (1-323AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: 3-alpha-HSD3, Chlordecone reductase homolog HAKRD, Dihydrodiol dehydrogenase 2, Dihydrodiol dehydrogenase/bile acid-binding protein, Trans-1, 2-dihydrobenzene-1, 2-diol dehydrogenase, Type III 3-alpha-hydroxysteroid dehydrogenase, AKR1C2, DDH2. UniProt: P52895. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7805