Learn More
Biomatik Corporation Aldh1a1 anti-Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08223
Encompass_Preferred
The Aldh1a1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: Recombinant mouse Retinal dehydrogenase 1 protein (2-501AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-2000. Alternative Names: Retinal dehydrogenase 1 (EC:1.2.1.36), RALDH 1, RalDH1, Aldh1a1, Ahd-2, Ahd2, Aldh1, ALDH-E1, ALHDII, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic. UniProt: P24549. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7807