Learn More
Biomatik Corporation ALDH5A1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08224
Encompass_Preferred
The ALDH5A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Succinate-semialdehyde dehydrogenase, mitochondrial protein (48-535AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Aldehyde dehydrogenase family 5 member A1, NAD (+)-dependent succinic semialdehyde dehydrogenase, SSADH, ALDH5A1. UniProt: P51649. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7808