Learn More
Biomatik Corporation ALOX15B anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08225
Encompass_Preferred
The ALOX15B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Arachidonate 15-lipoxygenase B protein (1-676AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: 15-LOX-B, 15-lipoxygenase 2, 15-LOX-2, Arachidonate 15-lipoxygenase type II, Linoleate 13-lipoxygenase 15-LOb, ALOX15B. UniProt: O15296. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7809