Learn More
Biomatik Corporation ALOX5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08226
Encompass_Preferred
The ALOX5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Arachidonate 5-lipoxygenase protein (2-149AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Arachidonate 5-lipoxygenase (EC:1.13.11.34), 5-LO, 5-lipoxygenase, ALOX5, LOG5. UniProt: P09917. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7810