Learn More
Biomatik Corporation AMBP anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507811
Encompass_Preferred
The AMBP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein AMBP protein (20-203AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Alternative name (s): Alpha-1 microglycoprotein Complex-forming glycoprotein heterogeneous in charge Bikunin EDC1 HI-30 Uronic-acid-rich protein AMBP HCP, ITIL. UniProt: P02760. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7811 Supplier Biomatik Corporation Supplier No. CAC08227
The AMBP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein AMBP protein (20-203AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Alternative name (s): Alpha-1 microglycoprotein Complex-forming glycoprotein heterogeneous in charge Bikunin EDC1 HI-30 Uronic-acid-rich protein AMBP HCP, ITIL. UniProt: P02760. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.