Learn More
Biomatik Corporation ANGPTL7 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08228
Encompass_Preferred
The ANGPTL7 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Angiopoietin-related protein 7 protein (27-296AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Angiopoietin-related protein 7, Angiopoietin-like factor, Angiopoietin-like protein 7, Cornea-derived transcript 6 protein, ANGPTL7, CDT6. UniProt: O43827. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7812