Learn More
Biomatik Corporation ANXA9 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08232
Encompass_Preferred
The ANXA9 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Annexin A9 protein (8-345AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Annexin XXXI, Annexin-31, Annexin-9, Pemphaxin, ANXA9, ANX31. UniProt: O76027. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7816