Learn More
Biomatik Corporation AOX1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08233
Encompass_Preferred
The AOX1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Aldehyde oxidase protein (236-421AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: AOX1, AO. UniProt: Q06278. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7817