Learn More
Biomatik Corporation AP1S3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507818
Encompass_Preferred
The AP1S3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human AP-1 complex subunit sigma-3 protein (1-104AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Adapter-related protein complex 1 sigma-1C subunit Adaptor protein complex AP-1 sigma-1C subunit Clathrin assembly protein complex 1 sigma-1C small chain Golgi adaptor HA1/AP1 adaptin sigma-1C subunit Sigma 1C subunit of AP-1 clathrin Sigma-adaptin 1. UniProt: Q96PC3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7818 Supplier Biomatik Corporation Supplier No. CAC08234
The AP1S3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human AP-1 complex subunit sigma-3 protein (1-104AA). Applications: ELISA, WB. Recommended dilution: WB:1:1000-1:5000. Alternative Names: Adapter-related protein complex 1 sigma-1C subunit Adaptor protein complex AP-1 sigma-1C subunit Clathrin assembly protein complex 1 sigma-1C small chain Golgi adaptor HA1/AP1 adaptin sigma-1C subunit Sigma 1C subunit of AP-1 clathrin Sigma-adaptin 1. UniProt: Q96PC3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.