Learn More
Biomatik Corporation APAF1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507819
Encompass_Preferred
The APAF1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Apoptotic protease-activating factor 1 protein (104-415AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:50-1:200, IF:1:50-1:500. Alternative Names: Apoptotic protease-activating factor 1, APAF-1, APAF1, KIAA0413. UniProt: O14727. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7819 Supplier Biomatik Corporation Supplier No. CAC08235
The APAF1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Apoptotic protease-activating factor 1 protein (104-415AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:50-1:200, IF:1:50-1:500. Alternative Names: Apoptotic protease-activating factor 1, APAF-1, APAF1, KIAA0413. UniProt: O14727. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.