Learn More
Biomatik Corporation APLNR anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507821
Encompass_Preferred
The APLNR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Apelin receptor protein (309-380AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Apelin receptor, APLNR, AGTRL1, APJ, Angiotensin receptor-like 1, G-protein coupled receptor APJ, G-protein coupled receptor HG11. UniProt: P35414. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7821 Supplier Biomatik Corporation Supplier No. CAC08237
The APLNR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Apelin receptor protein (309-380AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Apelin receptor, APLNR, AGTRL1, APJ, Angiotensin receptor-like 1, G-protein coupled receptor APJ, G-protein coupled receptor HG11. UniProt: P35414. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.