Learn More
Biomatik Corporation APOD anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08240
Encompass_Preferred
The APOD Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Apolipoprotein D protein (21-189AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Apo-D, ApoD, APOD. UniProt: P05090. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7824