Learn More
Biomatik Corporation ARFIP2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC08241
Encompass_Preferred
The ARFIP2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Arfaptin-2 protein (20-295AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Arfaptin-2, ARFIP2, POR1, ADP-ribosylation factor-interacting protein 2, Partner of RAC1, Protein POR1. UniProt: P53365. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7825